Learn More
Medchemexpress LLC HY-N2318 10mg Medchemexpress, Podocarpic acid CAS:5947-49-9 Purity:>98%
SDP
Catalog No. 501933910
Encompass_Preferred
Medchemexpress, HY-N2318 10mg Podocarpic acid CAS:5947-49-9 Podocarpic acid is a natural product, which has the best all-round positive effect and acts as a novel TRPA1 activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3910 Supplier Medchemexpress LLC Supplier No. HYN231810MG
Medchemexpress, HY-N2318 10mg Podocarpic acid CAS:5947-49-9 Podocarpic acid is a natural product, which has the best all-round positive effect and acts as a novel TRPA1 activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.