Learn More
Medchemexpress LLC Podocarpic acid | 5947-49-9 | 5 MG
SDP
Catalog No. 5000420348
Encompass_Preferred
Podocarpic acid is a natural product that functions as a novel TRPA1 activator. It has a molecular weight of 274.35 and the formula C17H22O3. The product appears as a white to off-white solid.
  • Acts as a novel TRPA1 activator
  • Isolated from Plants, Annonaceae, Dasymaschalon rostratum Merr. et Chun
  • Available in solid form
  • Suitable for in vitro and in vivo studies
Catalog No. 50-004-20348 Supplier Medchemexpress LLC Supplier No. HYN23185MG
Add to Cart
Add to Cart
Podocarpic acid is a natural product that functions as a novel TRPA1 activator. It has a molecular weight of 274.35 and the formula C17H22O3. The product appears as a white to off-white solid.
  • Acts as a novel TRPA1 activator
  • Isolated from Plants, Annonaceae, Dasymaschalon rostratum Merr. et Chun
  • Available in solid form
  • Suitable for in vitro and in vivo studies