Learn More
Enzo Life Sciences Podophyllotoxin (100mg). CAS: 518-28-5
SDP
Supplier:  Enzo Life Sciences ALX630086M100
Encompass_Preferred
Potent inhibitor of microtubule assembly. Antineoplastic glucoside. Antitumor agent. DNA topoisomerase II inhibitor. Purity: ≥98% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in chloroform, 100 % ethanol, acetone or dichloromethane; insoluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-9614