Learn More
AdipoGen Podophyllotoxin
SDP
Catalog No. 502562684
Encompass_Preferred
Chemical. CAS 518-28-5. Formula C22H22O8. MW 414.4. Isolated from Podophyllum emodi rhizomes. Potent microtubule assembly inhibitor. Anticancer compound. Cell death inducer. DNA topoisomerase II inhibitor. Cell cycle inhibitor at the metaphase. Antiviral and antihelminthic.
Catalog No. 50-256-2684 Supplier AdipoGen Supplier No. AGCN20049M500
Chemical. CAS 518-28-5. Formula C22H22O8. MW 414.4. Isolated from Podophyllum emodi rhizomes. Potent microtubule assembly inhibitor. Anticancer compound. Cell death inducer. DNA topoisomerase II inhibitor. Cell cycle inhibitor at the metaphase. Antiviral and antihelminthic.