Learn More
AdipoGen Podophyllotoxin
SDP
Catalog No. 502562685
Encompass_Preferred
Chemical. CAS 518-28-5. Formula C22H22O8. MW 414.4. Isolated from Podophyllum emodi rhizomes. Potent microtubule assembly inhibitor. Anticancer compound. Cell death inducer. DNA topoisomerase II inhibitor. Cell cycle inhibitor at the metaphase. Antiviral and antihelminthic.
Catalog No. 50-256-2685 Supplier AdipoGen Supplier No. AGCN20049M100
Chemical. CAS 518-28-5. Formula C22H22O8. MW 414.4. Isolated from Podophyllum emodi rhizomes. Potent microtubule assembly inhibitor. Anticancer compound. Cell death inducer. DNA topoisomerase II inhibitor. Cell cycle inhibitor at the metaphase. Antiviral and antihelminthic.