Learn More
Apexbio Technology LLC Podophyllotoxin 518-28-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000089622
Encompass_Preferred
Podophyllotoxin (CAS 518-28-5) is a naturally occurring lignan that acts as a microtubule assembly inhibitor by binding to tubulin thereby disrupting mitotic spindle formation and inhibiting cell division This mechanism underlies its antineoplastic activity making it a valuable tool in studies of cell cycle regulation cytoskeletal dynamics and anticancer drug development Podophyllotoxin is widely used in biomedical research to investigate mechanisms of cytotoxicity and to screen for compounds targeting mitotic pathways The compound is supplied as 100 mg in a glass bottle or 50 mg in a poly bottle
Catalog No. 50-000-89622 Supplier Apexbio Technology LLC Supplier No. N17905.1
Podophyllotoxin (CAS 518-28-5) is a naturally occurring lignan that acts as a microtubule assembly inhibitor by binding to tubulin thereby disrupting mitotic spindle formation and inhibiting cell division This mechanism underlies its antineoplastic activity making it a valuable tool in studies of cell cycle regulation cytoskeletal dynamics and anticancer drug development Podophyllotoxin is widely used in biomedical research to investigate mechanisms of cytotoxicity and to screen for compounds targeting mitotic pathways The compound is supplied as 100 mg in a glass bottle or 50 mg in a poly bottle