Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Podophyllotoxin, 95%

Catalog No. AC228410010
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC228410010 1 g
AC228411000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC228410010 Supplier Thermo Scientific Chemicals Supplier No. 228410010
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 518-28-5
Molecular Formula C22H22O8
Molecular Weight (g/mol) 414.41
MDL Number MFCD00075290
InChI Key YJGVMLPVUAXIQN-XVVDYKMHSA-N
Synonym podophyllotoxin, podofilox, condylox, condyline, wartec, podophyllinic acid lactone, podophyllotoxin 7, --podophyllotoxin, warticon, podophyllum
PubChem CID 10607
ChEBI CHEBI:50305
IUPAC Name (5R,5aR,8aR,9R)-5-hydroxy-9-(3,4,5-trimethoxyphenyl)-5a,6,8a,9-tetrahydro-5H-[2]benzofuro[5,6-f][1,3]benzodioxol-8-one
SMILES COC1=CC(=CC(OC)=C1OC)[C@H]1[C@@H]2[C@H](COC2=O)[C@@H](O)C2=CC3=C(OCO3)C=C12

Specifications

Melting Point 178.0°C to 184.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 7663
Specific Rotation − 110.70
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone and acetic acid
Formula Weight 414.4
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Podophyllotoxin, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal in contact with skin.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
EINECSNumber 208-250-4
RTECSNumber LV2500000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.