Learn More
Medchemexpress LLC Podophyllotoxin glucoside | 16481-54-2 | 576.55 | 1 MG
SDP
Catalog No. 5000415683
Encompass_Preferred
Podophyllotoxin glucoside is a derivative of podophyllotoxin, exhibiting anti-tumor effects. It is suitable for research use and has been tested for cytotoxicity against human HeLa and KB cells.
  • Exhibits anti-tumor effects
  • Tested for cytotoxicity against human HeLa cells: ED50 of 21.8 μg/mL (after 4 days by trypan blue assay) and IC50 of 6.65 μM (after 72 hours by MTT assay)
  • Tested for cytotoxicity against human KB cells: IC50 of 9.25 μM (after 72 hours by MTT assay)
Catalog No. 50-004-15683 Supplier Medchemexpress LLC Supplier No. HYN19771MG
Podophyllotoxin glucoside is a derivative of podophyllotoxin, exhibiting anti-tumor effects. It is suitable for research use and has been tested for cytotoxicity against human HeLa and KB cells.
  • Exhibits anti-tumor effects
  • Tested for cytotoxicity against human HeLa cells: ED50 of 21.8 μg/mL (after 4 days by trypan blue assay) and IC50 of 6.65 μM (after 72 hours by MTT assay)
  • Tested for cytotoxicity against human KB cells: IC50 of 9.25 μM (after 72 hours by MTT assay)