Learn More
Diagnostic Biosystems PODOPLANIN Podoplanin (D2-40)
Catalog No. 502407485
Encompass_Preferred
Podoplanin identifies the ~38 kDa O-linked transmembrane sialoglycoprotein podoplanin, which is expressed in the endothelium of lymphatic capillaries, but not in the blood vasculature (1). Besides the expression in lymphatic endothelium, podoplanin is also found in a variety of other tissues, including mesothelial cells, reticular cells, follicular dendritic cells, ovarian and testicular germ cells (2). Results aid in the classification of lymphatic invasion of primary tumors.
Catalog No. 50-240-7485 Supplier Diagnostic Biosystems Supplier No. PDM558
Podoplanin identifies the ~38 kDa O-linked transmembrane sialoglycoprotein podoplanin, which is expressed in the endothelium of lymphatic capillaries, but not in the blood vasculature (1). Besides the expression in lymphatic endothelium, podoplanin is also found in a variety of other tissues, including mesothelial cells, reticular cells, follicular dendritic cells, ovarian and testicular germ cells (2). Results aid in the classification of lymphatic invasion of primary tumors.