Learn More
Cayman Chemical PodoplnInRabbItMoncLNal100uL
SDP
Catalog No. 502910576
Encompass_Preferred
A saikosaponin with anti-inflammatory and analgesic activities reduces basal PGE2 production in isolated rat peritoneal macrophages at 100 Ug/ml decreases acetic acid-induced writhing in SART mice at 5 mg/kg per day
Catalog No. 50-291-0576 Supplier Cayman Chemical Supplier No. 35810100 UL
A saikosaponin with anti-inflammatory and analgesic activities reduces basal PGE2 production in isolated rat peritoneal macrophages at 100 Ug/ml decreases acetic acid-induced writhing in SART mice at 5 mg/kg per day