Learn More
CELL GUIDANCE SYSTEMS LLC PODS® GFP, 25 ug
SDP
Catalog No. NC3963446
Encompass
PODS are sold by cargo weight. This is typically 0.5 - 1.05 of the total protein weight. The remaining 99-99.5% is the polyhedrin protein. This GFP product contains polyhedrin protein co-crystalized with Green Fluorescent Protein (GFP). GFP is a uniquely versatile biomarker and, encased into PODS® crystals, offers a simple way to visualize und localize PODS® crystals embedded in biomaterials, such as hydrogels and scaffolds, using fluorescence microscopy. PODS® GFP crystals can be excited at 488 nm and optimally detected at 510 nm, compatible with commonly available filter sets.
Catalog No. NC3963446 Supplier CELL GUIDANCE SYSTEMS LLC Supplier No. PP30325
PODS are sold by cargo weight. This is typically 0.5 - 1.05 of the total protein weight. The remaining 99-99.5% is the polyhedrin protein. This GFP product contains polyhedrin protein co-crystalized with Green Fluorescent Protein (GFP). GFP is a uniquely versatile biomarker and, encased into PODS® crystals, offers a simple way to visualize und localize PODS® crystals embedded in biomaterials, such as hydrogels and scaffolds, using fluorescence microscopy. PODS® GFP crystals can be excited at 488 nm and optimally detected at 510 nm, compatible with commonly available filter sets.