Learn More
CELL GUIDANCE SYSTEMS LLC PODS® mCherry, 25 ug
SDP
Catalog No. NC3963451
Encompass
The product contains polyhedrin protein co-crystalized with mCherry. mCherry is a uniquely versatile biomarker and, encased into PODS® crystals, offers a simple way to visualize und localize PODS® crystals embedded in biomaterials, such as hydrogels and scaffolds, using fluorescence microscopy. PODS® mCherry crystals can be excited at 587 nm and optimally detected at 610 nm, compatible with commonly available filter sets.
Catalog No. NC3963451 Supplier CELL GUIDANCE SYSTEMS LLC Supplier No. PP30425
The product contains polyhedrin protein co-crystalized with mCherry. mCherry is a uniquely versatile biomarker and, encased into PODS® crystals, offers a simple way to visualize und localize PODS® crystals embedded in biomaterials, such as hydrogels and scaffolds, using fluorescence microscopy. PODS® mCherry crystals can be excited at 587 nm and optimally detected at 610 nm, compatible with commonly available filter sets.