Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 5420MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IP, WB, IHC. Conjugate/Tag: Unconjugated. Clone: 3D3. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: HeLa, Raji, Jurkat cells, Angiosarcoma, Breast, Prostate, Liver, Pancreatic, Kidney. Cellular Staining: Cell surface, cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: A recombinant protein fragment containing the intracellular, transmembrane,, part of the extracellular domain of human podocalyxin. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 165-170kDa.Chromosome: 7q32.3. Gene Symbol: PODXL. Gene ID: 5420. Ensemble: ENSG00000128567. Uniprot: NP_005388.
Catalog No. 50-188-5061