Learn More
Aviva Systems Biology Corp PODXL2 Antibody - N-terminal region (ARP47187_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47187_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PODXL2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PODXL2. Alternate names: PODLX2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-287