Learn More
Aviva Systems Biology Corp POFUT1 Antibody - middle region (ARP59441_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59441_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POFUT1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human POFUT1. Alternate names: FUT12, KIAA0180, MGC2482, O-FUT, O-Fuc-T, O-FucT-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-697