Learn More
Aviva Systems Biology Corp POFUT2 Antibody - C-terminal region (ARP45854_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45854_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against POFUT2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human POFUT2. Alternate names: C21orf80, FUT13ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-59152