Learn More
Aviva Systems Biology Corp POGK Antibody - middle region (ARP39288_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39288_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POGK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human POGK. Alternate names: BASS2, KIAA1513, KIAA15131, LST003, KRBOX2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56973