Learn More
ALPAQUA ENGINEERING LLC POGO CHILLER BLOCK
Catalog No. NC0591703
Encompass
POGO is made of solid aluminum to help keep your reagents cool while on automation decks. It is compatible with many common screw cap vials and micro-centrifuge tubes.
Catalog No. NC0591703 Supplier ALPAQUA ENGINEERING LLC Supplier No. A000020
POGO is made of solid aluminum to help keep your reagents cool while on automation decks. It is compatible with many common screw cap vials and micro-centrifuge tubes.