Learn More
Aviva Systems Biology Corp POGZ Antibody - middle region (ARP32982_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32982_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POGZ. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human POGZ. Alternate names: KIAA0461, MGC71543, SUHW5, ZNF280E, ZNF635, ZNF635mApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54842