Learn More
Aviva Systems Biology Corp POGZ Antibody - C-terminal region (ARP39173_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39173_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POGZ. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human POGZ. Alternate names: KIAA0461, MGC71543, SUHW5, ZNF280E, ZNF635, ZNF635mApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-785