Learn More
ICU Medical Point-Lok™ Needle Protection Device
Catalog No. 22004139
Encompass
Change view
Click to view available options
Quantity:
1000 Devices
500 Devices
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-004-139 1000 Devices
22-004-129 500 Devices
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-004-139 Supplier ICU Medical Supplier No. 4139
Add to Cart
Add to Cart
Point-Lok™ devices provide effective protection for needles from 16-30 g that do not have an integral engineered sharps injury protection (ESIP) device. It is available separately, or can be packaged in Portex™ Arterial Blood Sampling kits, pain management and other procedural trays.
  • Stand-alone needle protection device designed to lock onto and secure a contaminated needle.
  • Meets the Federal Register definition of an Engineering Control
Specifications
Product Type Needle Protection Device
Quantity 1000 Devices
For Use With (Equipment) Needles and Stylets 16 to 30G