Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Pokeweed mitogen native Protein (Crude) from pokeweeds

Catalog No. 89952541 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-952-541 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-952-541 Supplier Abnova™ Supplier No. P5356
Add to Cart
Add to Cart

Used for Enzyme, SDS-PAGE

Specifications

For Use With (Application) Enzyme Activity, SDS-PAGE
Name Mitogen (Pokeweed) Native Protein
Preparation Method Native protein purified from pokeweeds
Quantity 5 mg
Storage Requirements Store at -20°C on dry atmosphere.. After reconstitution with distilled water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name MITOGEN
Biological Activity Shows agglutination when tested with human red blood cells.
Species Plants
Protein Tag None
Expression System Native protein purified from pokeweeds
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.