Learn More
New England Biolabs, Inc. Poly (U) Polymerase – 60 units
SDP
Catalog No. 501959357
Encompass
Poly(U) Polymerase catalyzes the template independent addition of UMP from UTP or AMP from ATP to the 3' end of RNA.
Catalog No. 50-195-9357 Supplier New England Biolabs, Inc. Supplier No. M0337S
Poly(U) Polymerase catalyzes the template independent addition of UMP from UTP or AMP from ATP to the 3' end of RNA.