Learn More
Medchemexpress LLC HY-B0729 10mg Medchemexpress, Polaprezinc CAS:107667-60-7 Purity:>98%
SDP
Catalog No. 502082650
Encompass_Preferred
Medchemexpress, HY-B0729 10mg Polaprezinc CAS:107667-60-7 Polaprezinc is an orally bioavailable chelate composed of zinc and L-carnosine, with potential gastroprotective, anti-oxidant, anti-ulcer and anti-inflammatory activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2650 Supplier Medchemexpress LLC Supplier No. HYB072910MG
Medchemexpress, HY-B0729 10mg Polaprezinc CAS:107667-60-7 Polaprezinc is an orally bioavailable chelate composed of zinc and L-carnosine, with potential gastroprotective, anti-oxidant, anti-ulcer and anti-inflammatory activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.