Learn More
Cole-Parmer Argos Technologies Polarsafe EVA Foam Rectangular Ice Pan with Lid, 9 L, Blue; 1/EA
Catalog No. 501960588 Shop All Cole-Parmer Products
Encompass_Preferred
These EVA foam pans are ideally suited to benchtop chilling applications. They will not create puddles of condensed atmospheric vapor or leak cryogenic media onto your workspace. Ethylene vinyl acetate is compatible with ice, dry ice, liquid nitrogen, alcohol and saline solutions. Pans feature useful handles and convenient pour spouts for easy use and cleanup, even while wearing cryogenic gloves. Stackable construction ensures compact storage.
Catalog No. 50-196-0588 Supplier Cole-Parmer Supplier No. 0439376
These EVA foam pans are ideally suited to benchtop chilling applications. They will not create puddles of condensed atmospheric vapor or leak cryogenic media onto your workspace. Ethylene vinyl acetate is compatible with ice, dry ice, liquid nitrogen, alcohol and saline solutions. Pans feature useful handles and convenient pour spouts for easy use and cleanup, even while wearing cryogenic gloves. Stackable construction ensures compact storage.