Learn More
Avanti Polar Lipids 1,2-dioleoyl-sn-glycero-3-phospho-(1'-rac-glycerol) (sodium salt), 67254-28-8, MFCD00070069, 25mg
Catalog No. NC1122775
Encompass
Molecular Formula: C42H78O10PNa, Formula Weight: 797.026, Purity >99%.
Catalog No. NC1122775 Supplier Avanti Polar Lipids Supplier No. 840475C25MG
Molecular Formula: C42H78O10PNa, Formula Weight: 797.026, Purity >99%.