Learn More
Cayman Chemical Polar Lipid Mixture (egg, ovine), 1 mL
SDP
Catalog No. 502988269
Encompass_Preferred
A mixture of polar lipids containing cholesterol and both hydroxy- and non-hydroxy phosphatidylethanolamines, phosphatidylcholines, and lysophosphatidylcholines; intended for use as a reference standard for the detection of these polar lipids in various sample types by TLC or LC-MS
Catalog No. 50-298-8269 Supplier Cayman Chemical Supplier No. 293741 ML
A mixture of polar lipids containing cholesterol and both hydroxy- and non-hydroxy phosphatidylethanolamines, phosphatidylcholines, and lysophosphatidylcholines; intended for use as a reference standard for the detection of these polar lipids in various sample types by TLC or LC-MS