Learn More
Analytical Sales & Services Polar Phase Cyano SPE 96-Well Plate, 1.0mL/50mg Bed
Catalog No. 5000807012
Encompass
CN (cyanol) SPE, 1mL 96 Well Filter Plate, 50mg per well
Catalog No. 50-008-07012 Supplier Analytical Sales & Services Supplier No. 9610CN50
CN (cyanol) SPE, 1mL 96 Well Filter Plate, 50mg per well