Learn More
Analytical Sales & Services Polar Phase Diol SPE 96-Well Plate, 1.0mL/100mg Bed
Catalog No. 5000804438
Encompass
Diol SPE, 1mL 96 Well Filter Plate, 100mg per well
Catalog No. 50-008-04438 Supplier Analytical Sales & Services Supplier No. 9610DI100
Diol SPE, 1mL 96 Well Filter Plate, 100mg per well