Learn More
Analytical Sales & Services Polar Phase Diol SPE 96-Well Plate, 1.0mL/25mg Bed
Catalog No. 5000806823
Encompass
Diol SPE, 1mL 96 Well Filter Plate, 25mg per well
Catalog No. 50-008-06823 Supplier Analytical Sales & Services Supplier No. 9610DI25
Diol SPE, 1mL 96 Well Filter Plate, 25mg per well