Learn More
Analytical Sales & Services Polar Phase NH (amino) SPE 96-Well Plate, 1.0mL/25mg Bed
Catalog No. 5000806354
Encompass
NH (amino) SPE, 1mL 96 Well Filter Plate, 25mg per well
Catalog No. 50-008-06354 Supplier Analytical Sales & Services Supplier No. 9610NH25
Add to Cart
Add to Cart
NH (amino) SPE, 1mL 96 Well Filter Plate, 25mg per well