Learn More
Analytical Sales & Services Polar Phase NH (amino) SPE 96-Well Plate, 1.0mL/50mg Bed
Catalog No. 5000806355
Encompass
NH (amino) SPE, 1mL 96 Well Filter Plate, 50mg per well
Catalog No. 50-008-06355 Supplier Analytical Sales & Services Supplier No. 9610NH50
NH (amino) SPE, 1mL 96 Well Filter Plate, 50mg per well