Learn More
Analytical Sales & Services Polar Phase Silica SPE 96-Well Plate, 1.0mL/50mg Bed
Catalog No. 5000806788
Encompass
Silica SPE, 1mL 96 Well Filter Plate, 50mg per well
Catalog No. 50-008-06788 Supplier Analytical Sales & Services Supplier No. 9610SI50
Silica SPE, 1mL 96 Well Filter Plate, 50mg per well