Learn More
WELCH MATERIALS INC Ultisil Polar RP, 10µm, 50×250mm
Catalog No. NC2052702
Encompass_Preferred
Prep Column, Ultisil Polar RP, 10µm, 50×250mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, L60, Carbon Loading: 18%, Surface Area: 320m2/g, endcapped.
Catalog No. NC2052702 Supplier WELCH MATERIALS INC Supplier No. H0262131210
Add to Cart
Add to Cart
Request a Quote
Prep Column, Ultisil Polar RP, 10µm, 50×250mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, L60, Carbon Loading: 18%, Surface Area: 320m2/g, endcapped.