Learn More
Solstice Polar Silica-Based SPE Columns, Aminopropyl (NH2), 300 mg/3 mL
Catalog No. 6002751 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
50
Catalog No. Quantity
600-27-51 50
1 options
Catalog No. 600-27-51 Supplier Solstice Supplier No. 9073
Add to Cart
Add to Cart
Medical-grade polypropylene with PE frits
  • All columns are normal phase and should be preconditioned with a non-polar solvent with the elution solvent being polar
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50