Learn More
Solstice Polar Silica-Based SPE Columns, Silica (SI), 200 mg/3 mL
Shop All Solstice Research Chemicals ProductsDescription
All columns are normal phase and should be preconditioned with a non-polar solvent with the elution solvent being polar
Specifications
Specifications
| Quantity | 50 |