Learn More
Solstice Polar Silica-Based SPE Columns, Silica (SI), 500 mg/6 mL
Catalog No. 6002881 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
30
Catalog No. Quantity
600-28-81 30
1 options
Catalog No. 600-28-81 Supplier Solstice Supplier No. 9056
Add to Cart
Add to Cart
Medical-grade polypropylene with PE frits
  • All columns are normal phase and should be preconditioned with a non-polar solvent with the elution solvent being polar
TRUSTED_SUSTAINABILITY
Specifications
Quantity 30
WARNING: Reproductive Harm - www.P65Warnings.ca.gov