Learn More
Polar Silica-Based SPE Columns, Silica (SI), Medical-grade polypropylene with PE frits, 500 mg/25 mL, Solstice
Shop All Solstice Research Chemicals ProductsDescription
All columns are normal phase and should be preconditioned with a non-polar solvent with the elution solvent being polar
Specifications
Specifications
| Quantity | 100 Pc. |
| Packaging | Fiberboard Box |
| Grade | Analytical |
| Chemical Name or Material | Polar Silica-Based SPE Columns, Silica (SI), Medical-grade polypropylene with PE frits, 500 mg/25 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.