Learn More
Azzota Corp Polarimeter Tube 200mm 16ml
Catalog No. 5000887238
Encompass_Preferred
Polarimeter tube - Constant Temperature model Pathlength 200mm /- 1% Diameter 14mm Volume 16ml
Catalog No. 50-008-87238 Supplier Azzota Corp Supplier No. APTT200
Polarimeter tube - Constant Temperature model Pathlength 200mm /- 1% Diameter 14mm Volume 16ml