Learn More
Anton Paar HASTELLOY C-276, LUER ADAPTER, FOR PELTIER, 100 mm for MCP 150
Catalog No. NC1710887
Encompass_Preferred
100 mm hastelloy sample cell (polarimeter tube) with wireless Toolmaster function which transfers temperature and path length to the MCP 150.Filling volume approximately 2 mL.Connection to a syringe through Luer filling ports.Suitable for use with Anton Paar polarimeter MCP 150 item NC1779945
Catalog No. NC1710887 Supplier Anton Paar Supplier No. 109920
100 mm hastelloy sample cell (polarimeter tube) with wireless Toolmaster function which transfers temperature and path length to the MCP 150.Filling volume approximately 2 mL.Connection to a syringe through Luer filling ports.Suitable for use with Anton Paar polarimeter MCP 150 item NC1779945