Learn More
Anton Paar STAINLESS STEEL, LUER ADAPTER, FOR PELTIER, 100 mm for MCP 150
Catalog No. NC1779947
Encompass_Preferred
100 mm stainless steel sample cell (polarimeter tube) with wireless Toolmaster function which transfers temperature and path length to the MCP 150.Filling volume approximately 2 mL.Connection to a syringe through Luer filling ports.Suitable for use with Anton Paar polarimeter MCP 150 item NC1779945
Catalog No. NC1779947 Supplier Anton Paar Supplier No. 104135
100 mm stainless steel sample cell (polarimeter tube) with wireless Toolmaster function which transfers temperature and path length to the MCP 150.Filling volume approximately 2 mL.Connection to a syringe through Luer filling ports.Suitable for use with Anton Paar polarimeter MCP 150 item NC1779945