Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Polarization Demonstrator
GREENER_CHOICESCI_ED

Catalog No. S160132 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
9 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S160132 9 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S160132 Supplier Supertek™ Supplier No. PH30642
Add to Cart
Add to Cart
This item is not returnable. View return policy

Demonstrate stress patterns in solids with this new and valuable Supertek™ Polarization Demonstrator.

  • When acrylic pieces are introduced between the filters and when pressure is applied, the resulting stress patterns are observed.
  • Size: 7.08 L x 4.52 W x 2.36 H in.
  • Includes instructions.

Specifications

Grade 9 to Undergraduate
Format Kit
Includes Plastic base, 2 polarizing filters mounted on graduated frames, and 4 clear acrylic pieces of different shapes.
Languages English
Subject Physics - Matter and Its Properties
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.