Learn More
Vernier™ Polarizer/Analyzer for Optics Expansion Kit
SDPSCI_ED
Catalog No. S16002ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With: Optics Expansion Kit
Catalog No.
S16002ND For Use With: Optics Expansion Kit
1 options
Catalog No. S16002ND Supplier Vernier™ Supplier No. PAKOEK
Add to Cart
Add to Cart
Includes
Adjustable Analyzer (2), Adjustable Analyzer for Rotary Motion Sensor (1), Mounting Screws for Rotary Motion Sensor (2), Belt for Rotary Motion Sensor (1)
The Polarizer/Analyzer Set extends the Optics Expansion Kit to allow students to study polarization of light. Using a Rotary Motion Sensor to record analyzer angle, Malus's Law experiments are easy, detailed, and accurate.
Polarizer passes vertically polarized light. Adjustable Analyzer for Rotary Motion Sensor includes a mount for the Vernier Rotary Motion Sensor to record the analyzer angle. Adjustable Analyzer includes an angle scale for manual reading of the polarizer angle.
Warranty and Services
5-year limited warranty
Specifications
For Use With (Application) Educational use only
For Use With (Equipment) Optics Expansion Kit
Includes 2 Adjustable Analyzers, 1 Adjustable Analyzer for Rotary Motion Sensor, 2 Mounting Screws for Rotary Motion Sensor, and 1 Belt for Rotary Motion Sensor