Description
- Epi-Illuminator offers a powerful 50 W halogen light source
- Built-in field and aperture diaphragms help to optimize image quality by reducing stray light and increasing contrast
- Simple polarization set enables useful 'POL contrast' and helps to minimize reflections, especially in low power objectives
Specifications
Specifications
| Type | Polarizer |
| For Use With (Application) | Minimizes Reflections |
| For Use With (Equipment) | BA310MET Series and Panthera TEC MET Series |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov