Learn More
Perkin Elmer US LLC Polarizer for 80Spec - ZnSe
Catalog No. 502113275
Encompass_Preferred
The PIKE 80Spec (L1272248) is a grazing angle specular reflection accessory that measures thin films and monolayers. Features include: A fixed 80° angle of incidence Polarizer mounts for optional IR polarizers (L1272376, L1272243) Gold-coated reflective optics for highest energy throughput It is compatible with Spectrum One/100/400/65, Spectrum Two, and Spectrum 3/Frontier spectrometers.
Catalog No. 50-211-3275 Supplier Perkin Elmer US LLC Supplier No. L1272376
The PIKE 80Spec (L1272248) is a grazing angle specular reflection accessory that measures thin films and monolayers. Features include: A fixed 80° angle of incidence Polarizer mounts for optional IR polarizers (L1272376, L1272243) Gold-coated reflective optics for highest energy throughput It is compatible with Spectrum One/100/400/65, Spectrum Two, and Spectrum 3/Frontier spectrometers.