Learn More
Motic Polarizing Equipment
Catalog No. 01550020 Shop All Motic Group Products
Catalog No. 01-550-020 Supplier Motic Supplier No. 1101000600641
Add to Cart
Add to Cart
B301K is a one-piece rotary polarizing equipment designed for the N2GG stand.
  • One-piece rotary design for convenient operation and adjustment.
  • Polarizing capability for observing specimens with polarization-dependent optical properties.
  • Rotary adjustment allows the polarization orientation to be changed during observation.
  • Designed as a stereo microscope accessory, including use within Motic SMZ-series configurations.
  • Compact integrated construction, avoiding the need for a separate multi-piece polarizing setup.
  • Useful for enhancing contrast and reducing unwanted reflections/glare when examining suitable materials under polarized illumination.