Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ PolarScreen™ Progesterone Receptor Competitor Assay Kit, Red

Catalog No. A15906
Encompass_Preferred
Change view
Click to view available options
Quantity:
400 x 20 μL assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15906 400 x 20 μL assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A15906 Supplier Thermo Scientific™ Supplier No. A15906
Add to Cart
Add to Cart

The PolarScreen™ Progesterone Receptor Competitor Assay Kit, Red, provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential progesterone receptor (PR) ligands.

The PolarScreen™ Progesterone Receptor Competitor Assay Kit, Red, provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential progesterone receptor (PR) ligands. The kit uses insect cell-expressed, human progesterone receptor ligand binding domain tagged with glutathione-S-transferase (GST) (also available separately) and a novel, high-affinity, fluorescent progesterone ligand (Fluormone™ PL Red) in a homogenous, mix-and-read assay format.

This kit produces a red color read-out; a 'green' kit is also available. Both kits suitably determine the IC50s of competitive compounds. Color choice depends on the spectral properties of the compounds to be screened and the available instrumentation. For additional information or assistance contact us at drugdiscoverytech@lifetech.com.

The PolarScreen Progesterone Receptor Competitor Assay is:

  • Complete & ready-to-use out of the box—all reagents included, just add your test compounds
  • High throughput compatible—fluorescence polarization assays in a 384-well plate
  • Convenient—components available in bulk sizes for larger screening needs

How the Assay Works

An N-terminal fusion of glutathione transferase to the ligand-binding domain of the human progesterone receptor (PR-LBD(GST)) is added to a fluorescently-tagged progesterone ligand (Fluormone PL Red) in the presence of competitor test compounds in microwell plates. The presence of effective competitors prevents the formation of a PL Red/PR-LBD(GST) complex resulting in a decrease of the polarization value. The shift in polarization value in the presence of test compounds is used to determine relative affinity of test compounds for PR-LBD(GST).

Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 400 x 20 μL assays
Content And Storage • Progesterone Receptor Ligand Binding Domain Recombinant Protein, 1 tube (store at -68°C to -85°C)
• Fluormone PL Red Tracer, 1 tube (-5°C to -30°C)
• PR Red Screening Buffer, 2 bottles (-5°C to -30°C)
• DTT, 1 tube (-5°C to -30°C)
Product Line PolarScreen
For Use With (Application) Competitive Binding Assay, Nuclear Receptor Assay
Ligand PR
Product Type Assay Kit
No. of Assays 400 x 20 μL Assays
What are the common issues encountered with FP assays?

-No assay window: Check that your instrument is designed to do FP experiments.
-Poor discrimination between the minimum and maximum mP Control (both include the receptor): This may be an indication of the quality of the receptor. Do not make single use aliquots of the receptor or store diluted.
-The type of microplate is critical: Please see if the protocol specifies untreated polystyrene or NBS-coated plates. If the plate binds the fluorescent substrate, it cannot freely rotate and the mP reading will always be high. White plates cannot be used for FP assays. Avoid the use of silanized pipette tips and vials.
-The “No Receptor Control' (free Fluormone/Tracer) should have much higher RFU values compared to buffer alone. If not, there is a problem with the instrument setup or less commonly with the Fluormone/Tracer.
-If the free Fluormone/Tracer mP value is greater than 100 mP or negative, please refer to the user manual for your instrument and reset the G factor.

What is the best type of microplate reader for fluorescence polarization (FP) assays?

For FP assays, plate readers must specifically have FP built in. Please see our instrument compatibility portal (http://www.thermofisher.com/us/en/home/industrial/pharma-biopharma/drug-discovery-development/dd-misc/instrument-compatibility-portal.html).

How can I test the fluorescence polarization (FP) settings of my microplate reader before doing the assay?

We offer a FP One-Step Reference Kit (https://www.thermofisher.com/order/catalog/product/P3088) to detect in green (Ex/Em = 488/535 nm) and red (Ex/Em = 525/590 nm) emission ranges, suitable for the PolarScreen Green and Red Assays. The alternative is to perform the Controls in the respective assay kit prior to performing the actual assay using test compounds.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.