Learn More
Selleck Chemical LLC Polatuzumab vedotin-piiq-1MG
SDP
Catalog No. 503135649
Encompass_Preferred
Polatuzumab vedotin-piiq (polatuzumab vedotin DCDS4501A RG7596) is an antibody-drug conjugate (ADC) composed of a monoclonal antibody against CD79b (a B cell receptor component) and the anti-mitotic cytotoxic agent monomethyl auristatin (MMAE) It is used for the treatment of relapsed/refractory diffuse large B-cell lymphoma (DLBCL)
Catalog No. 50-313-5649 Supplier Selleck Chemical LLC Supplier No. D40121MG
Polatuzumab vedotin-piiq (polatuzumab vedotin DCDS4501A RG7596) is an antibody-drug conjugate (ADC) composed of a monoclonal antibody against CD79b (a B cell receptor component) and the anti-mitotic cytotoxic agent monomethyl auristatin (MMAE) It is used for the treatment of relapsed/refractory diffuse large B-cell lymphoma (DLBCL)