Learn More
Aviva Systems Biology Corp POLDIP2 Antibody - C-terminal region (ARP65809_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65809_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POLDIP2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human POLDIP2. Alternate names: DKFZp586F1524, PDIP38, POLD4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-575