Learn More
Aviva Systems Biology Corp POLDIP3 Antibody - C-terminal region (ARP41047_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41047_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POLDIP3. It was validated on Western Blot. . Alternate names: KIAA1649, PDIP46, SKARApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rat
Catalog No. 50-284-908